CAS RN 57757-57-0
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 3-({3-[(2,5-dioxopyrrolidin-1-yl)oxy]-3-oxopropyl}disulfanyl)propanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 3-({3-[(2,5-dioxopyrrolidin-1-yl)oxy]-3-oxopropyl}disulfanyl)propanoate
Molecular Weight (g/mol):
404.41
Molecular Formula:
C14H16N2O8S2
InChi Key:
FXYPGCIGRDZWNR-UHFFFAOYSA-N
SMILES:
O=C(CCSSCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
Di(N-succinimidyl) 3,3'-Dithiodipropionate 97.0+%, TCI America™
CAS: 57757-57-0 Molecular Formula: C14H16N2O8S2 Molecular Weight (g/mol): 404.408 MDL Number: MFCD00042045 InChI Key: FXYPGCIGRDZWNR-UHFFFAOYSA-N PubChem CID: 93313 IUPAC Name: (2,5-dioxopyrrolidin-1-yl) 3-[[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]disulfanyl]propanoate SMILES: C1CC(=O)N(C1=O)OC(=O)CCSSCCC(=O)ON2C(=O)CCC2=O