missing translation for 'onlineSavingsMsg'
Learn More

Di-n-pentyl sulfide, 97%

CAS: 872-10-6 | C10H22S | 174.35 g/mol

$50.32 - $168.07

Chemical Identifiers

CAS 872-10-6
Molecular Formula C10H22S
Molecular Weight (g/mol) 174.35
MDL Number MFCD00027275
InChI Key JOZDADPMWLVEJK-UHFFFAOYSA-N
Synonym pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide
PubChem CID 13382
IUPAC Name 1-pentylsulfanylpentane
SMILES CCCCCSCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0464906
View Documents
Thermo Scientific Chemicals
L0464906
5 g
Each for $50.32
Only null left
 
AAL0464914
View Documents
Thermo Scientific Chemicals
L0464914
25 g
Each for $168.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

872-10-6
174.35
JOZDADPMWLVEJK-UHFFFAOYSA-N
13382
CCCCCSCCCCC
C10H22S
MFCD00027275
pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide
1-pentylsulfanylpentane

Specifications

872-10-6
0.84
85°C (185°F)
C10H22S
CH3(CH2)4S(CH2)4CH3
5 g
pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide
CCCCCSCCCCC
174.35
174.35
Di-n-pentyl sulfide
-51°C
228°C to 230°C
Stench
1.456
MFCD00027275
1698031
JOZDADPMWLVEJK-UHFFFAOYSA-N
1-pentylsulfanylpentane
13382
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

EINECSNumber : 212-820-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only