missing translation for 'onlineSavingsMsg'
Learn More

Di-n-pentyl sulfide, 97%

Catalog No. AAL0464906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0464906 5 g
AAL0464914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0464906 Supplier Thermo Scientific Chemicals Supplier No. L0464906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 872-10-6 | C10H22S | 174.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 872-10-6
Molecular Formula C10H22S
Molecular Weight (g/mol) 174.35
MDL Number MFCD00027275
InChI Key JOZDADPMWLVEJK-UHFFFAOYSA-N
Synonym pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide
PubChem CID 13382
IUPAC Name 1-pentylsulfanylpentane
SMILES CCCCCSCCCCC

Specifications

Melting Point -51°C
Density 0.84
Boiling Point 228°C to 230°C
Flash Point 85°C (185°F)
Odor Stench
Refractive Index 1.456
Linear Formula CH3(CH2)4S(CH2)4CH3
Quantity 5 g
Beilstein 1698031
Formula Weight 174.35
Percent Purity 97%
Chemical Name or Material Di-n-pentyl sulfide
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-820-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only