missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl sulfide, 97%

Catalog No. p-7026362
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2232514 25 g
AAB2232522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2232514 Supplier Thermo Scientific Chemicals Supplier No. B2232514
Only null left

CAS: 6294-31-1 | C12H26S | 202.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6294-31-1
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00009528
InChI Key LHNRHYOMDUJLLM-UHFFFAOYSA-N
Synonym hexyl sulfide, hexane, 1,1'-thiobis, dihexyl sulfide, di-n-hexyl sulfide, 7-thiatridecane, dihexyl sulphide, n-hexyl sulfide, di-n-hexylsulfide, hexylthiohexane, normal-hexyl sulfide
PubChem CID 80517
IUPAC Name 1-hexylsulfanylhexane
SMILES CCCCCCSCCCCCC

Specifications

Density 0.849
Boiling Point 260°C
Flash Point 116°C (241°F)
Odor Stench
Refractive Index 1.4587
Quantity 25 g
UN Number UN3334
Beilstein 1738333
Formula Weight 202.4
Percent Purity 97%
Chemical Name or Material Di-n-hexyl sulfide
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber 228-556-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only