accessibility menu, dialog, popup

UserName

H3CSCH3

CAS RN 6294-31-1

IUPAC Name: 1-(hexylsulfanyl)hexane
Synonyms: di-n-hexyl sulfide
Molecular Weight (g/mol): 202.4
Molecular Formula: C12H26S
InChi Key: LHNRHYOMDUJLLM-UHFFFAOYSA-N
SMILES: CCCCCCSCCCCCC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)