missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals

$119.33 - $1851.81

Chemical Identifiers

CAS 84-75-3
Molecular Formula C20H30O4
Molecular Weight (g/mol) 334.46
MDL Number MFCD00043720
InChI Key KCXZNSGUUQJJTR-UHFFFAOYSA-N
Synonym dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate
PubChem CID 6786
ChEBI CHEBI:34678
IUPAC Name dihexyl benzene-1,2-dicarboxylate
SMILES CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1259309
View Documents
Thermo Scientific Chemicals
A1259309
10 g
Each for $119.33
Only null left
 
AAA1259318
View Documents
Thermo Scientific Chemicals
A1259318
50 g
Each for $441.33
Only null left
 
AAA1259330
View Documents
Thermo Scientific Chemicals
A1259330
250 g
Each for $1,851.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalates are by far the most widely used plasticizers, primarily to make soft and flexible polyvinyl chloride (PVC) for the applications in the industry of automotive, building & construction material, cable, flooring, medical device and toys. Minor quantity of phthalates are used in adhesives, caulk, sealants, paint to improve work performance. Small molecule phthalates are used as solvents in perfumes to provide longer linger longer and in nail polish to prevent chipping.

Solubility
Soluble in water (0.24 mg/L).

Notes
Store at 4°C. Avoid excessive heat and light. Reactive towards oxidizing agents.
Specifications

Chemical Identifiers

84-75-3
334.46
KCXZNSGUUQJJTR-UHFFFAOYSA-N
6786
dihexyl benzene-1,2-dicarboxylate
C20H30O4
MFCD00043720
dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate
CHEBI:34678
CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC

Specifications

84-75-3
185°C to 187°C (0.5 mmHg)
1.487
10 g
dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate
KCXZNSGUUQJJTR-UHFFFAOYSA-N
dihexyl benzene-1,2-dicarboxylate
6786
334.46
Di-n-hexyl phthalate
1.01
C20H30O4
MFCD00043720
UN3082
Soluble in water (0.24mg/L).
CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC
334.46
CHEBI:34678
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360FD

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 201-559-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only