missing translation for 'onlineSavingsMsg'
Learn More

Di-2-thienyl sulfide, 97%

Catalog No. p-7031983
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3328403 1 g
AAH3328406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3328403 Supplier Thermo Scientific Chemicals Supplier No. H3328403
Only null left

CAS: 3988-99-6 | C8H6S3 | 198.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3988-99-6
Molecular Formula C8H6S3
Molecular Weight (g/mol) 198.32
MDL Number MFCD00052047
InChI Key AKYIWBKPINOXJY-UHFFFAOYSA-N
Synonym 2-2-thienylthio thiophene, thiophene, 2,2'-thiobis, thiophene,2,2'-thiobis, 2-2-thienylsulfanyl thiophene, 2-thiophen-2-ylsulfanyl thiophene, 2-thienyl sulfide, di-2-thienyl sulfide, 2,2'-thiobisthiophene, di 2-thienyl sulphide
PubChem CID 138094
IUPAC Name 2-thiophen-2-ylsulfanylthiophene
SMILES S(C1=CC=CS1)C1=CC=CS1

Specifications

Density 1.311
Boiling Point 102°C to 104°C (0.9 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.649
Quantity 1 g
Formula Weight 198.32
Percent Purity 97%
Chemical Name or Material Di-2-thienyl sulfide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only