accessibility menu, dialog, popup

UserName

CAS RN 3988-99-6

IUPAC Name: 2-(thiophen-2-ylsulfanyl)thiophene
Synonyms: 2-(thiophen-2-ylsulfanyl)thiophene
Molecular Weight (g/mol): 198.32
Molecular Formula: C8H6S3
InChi Key: AKYIWBKPINOXJY-UHFFFAOYSA-N
SMILES: S(C1=CC=CS1)C1=CC=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)