missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.282 N Standardized Solution, Spectrum™ Chemical

Catalog No. 18613191
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613191 4 L Amber Glass Bottle
18613189 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18613191 Supplier Spectrum Chemical Mfg Cor Supplier No. S1704LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Physical Form Liquid
Quantity 4 L
Grade Not Available
Concentration or Composition (by Analyte or Components) 0.282 N ± 0.001 N
Packaging Amber Glass Bottle
Chemical Name or Material Silver Nitrate, 0.282 N Standardized Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.