missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Monohydrate, Powder, USP, 97.5-102%, Spectrum™ Chemical
DE145, 77938-63-7, C6H12O6·H2O
$269.21 - $4,239.70
Chemical Identifiers
| CAS | 77938-63-7 |
|---|---|
| Molecular Formula | C6H14O7 |
| Molecular Weight (g/mol) | 198.17 |
| MDL Number | MFCD00149450 |
| InChI Key | SPFMQWBKVUQXJV-UHFFFAOYNA-N |
| IUPAC Name | 2,3,4,5,6-pentahydroxyhexanal hydrate |
| SMILES | O.OCC(O)C(O)C(O)C(O)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602121
|
Spectrum Chemical Mfg Cor
DE145100GM |
100 g | Poly Bottle |
N/A
|
|
||||
|
18602117
|
Spectrum Chemical Mfg Cor
DE145500GM |
500 g | Poly Bottle |
N/A
|
|
||||
|
18602118
|
Spectrum Chemical Mfg Cor
DE14525KG |
2.5 kg | Poly Pail |
N/A
|
|
||||
|
18602119
|
Spectrum Chemical Mfg Cor
DE14512KG |
12 kg | Poly Pail |
N/A
|
|
||||
|
18602120
|
Spectrum Chemical Mfg Cor
DE14545KGBL |
45 kg | Fiber Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Dextrose, Monohydrate, Powder, USP - All Spectrum Chemical USP products, including all USP dextrose products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 77938-63-7 | |
| 198.17 | |
| SPFMQWBKVUQXJV-UHFFFAOYNA-N | |
| O.OCC(O)C(O)C(O)C(O)C=O |
| C6H14O7 | |
| MFCD00149450 | |
| 2,3,4,5,6-pentahydroxyhexanal hydrate |
Specifications
| 77938-63-7 | |
| Poly Bottle | |
| MFCD00149450 | |
| SPFMQWBKVUQXJV-UHFFFAOYNA-N | |
| 2,3,4,5,6-pentahydroxyhexanal hydrate | |
| 97.5 to 102% |
| 100% | |
| C6H14O7 | |
| 100 g | |
| O.OCC(O)C(O)C(O)C(O)C=O | |
| 198.17 | |
| USP |