missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Anhydrous, EP, BP, JP, USP, 99.5%, Spectrum™ Chemical
DE140, 50-99-7, C6H12O6
$334.03 - $2071.86
Chemical Identifiers
| CAS | 50-99-7 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.16 |
| InChI Key | WQZGKKKJIJFFOK-UHFFFAOYNA-N |
| IUPAC Name | 6-(hydroxymethyl)oxane-2,3,4,5-tetrol |
| SMILES | OCC1OC(O)C(O)C(O)C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602113
|
Spectrum Chemical Mfg Cor
DE140500GM |
500 g | Poly Bottle |
Each for $334.03
|
|
||||
|
18602114
|
Spectrum Chemical Mfg Cor
DE14025KG |
2.5 kg | Poly Bottle |
Each for $477.23
|
|
||||
|
18602115
|
Spectrum Chemical Mfg Cor
DE14012KG |
12 kg | Poly Pail |
Each for $2,071.86
|
|
||||
Description
Spectrum™ Chemical Dextrose, Anhydrous, USP, EP, BP, JP - All Spectrum Chemical USP, EP, BP, JP products, including all USP dextrose products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 50-99-7 | |
| 180.16 | |
| 6-(hydroxymethyl)oxane-2,3,4,5-tetrol |
| C6H12O6 | |
| WQZGKKKJIJFFOK-UHFFFAOYNA-N | |
| OCC1OC(O)C(O)C(O)C1O |
Specifications
| 50-99-7 | |
| 0.1% | |
| Poly Bottle | |
| 500 g | |
| OCC1OC(O)C(O)C(O)C1O | |
| 180.16 | |
| Multi Compendia |
| 100% | |
| 1% | |
| C6H12O6 | |
| WQZGKKKJIJFFOK-UHFFFAOYNA-N | |
| 6-(hydroxymethyl)oxane-2,3,4,5-tetrol | |
| 99.5% |