missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose Anhydrous (Crystalline Powder/USP/EP/BP/JP), Fisher Chemical™
Supplier: Fisher Chemical D1912
Specifications
| Dextrose Anhydrous | |
| 146.1°C | |
| Poly Pail | |
| CH2OH(CHOH)4CHO | |
| 12 kg | |
| Pass Test (USP) | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| 180.156 | |
| CHEBI:42758 | |
| ≥99.5% (JP) | |
| Solid |
| 50-99-7 | |
| 5.9 | |
| C6H12O6 | |
| MFCD00148912 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
| 107526 | |
| 180.16 | |
| BP/EP/JP/USP |
Chemical Identifiers
| 50-99-7 | |
| 180.156 | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| 107526 | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
| C6H12O6 | |
| MFCD00148912 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| CHEBI:42758 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
Safety and Handling