missing translation for 'onlineSavingsMsg'
Learn More

Dextrose Anhydrous (Crystalline Powder/USP/EP/BP/JP), Fisher Chemical™

Catalog No. p-2363434
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Flint Glass
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D19212 2.5 kg Flint Glass
D1912 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19212 Supplier Thermo Fisher Scientific Supplier No. D19212
Only null left

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148912
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.1°C
pH 5.9
Packaging Flint Glass
Linear Formula CH2OH(CHOH)4CHO
Quantity 2.5 kg
Residual Solvents Pass Test (USP)
Formula Weight 180.16
Percent Purity ≥99.5% (JP)
Grade BP/EP/JP/USP
Physical Form Solid
Chemical Name or Material Dextrose Anhydrous
Show More Show Less