missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran Sulfate Sodium Salt, Powder, Spectrum™ Chemical
DE136, 9011-18-1
$1007.36 - $92316.89
Chemical Identifiers
| CAS | 9011-18-1 |
|---|---|
| Molecular Formula | (C6H7O5)m(C6H7O5)n |
| Molecular Weight (g/mol) | 40,000 |
| MDL Number | MFCD00081551 |
| InChI Key | BLFLLBZGZJTVJG-UHFFFAOYSA-N |
| IUPAC Name | Dextran sulfate sodium salt |
| SMILES | *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602079
|
Spectrum Chemical Mfg Cor
DE13625GM |
25 g | Poly Bottle |
Each for $1,007.36
|
|
||||
|
18602080
|
Spectrum Chemical Mfg Cor
DE136100GM |
100 g | Poly Bottle |
Each for $3,214.18
|
|
||||
|
18602082
|
Spectrum Chemical Mfg Cor
DE1361KGBL |
1 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602081
|
Spectrum Chemical Mfg Cor
DE13610KGBL |
10 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Dextran Sulfate Sodium Salt, Powder is a sodium salt of the sulfuric acid ester of Dextran with an average molecular weight of 40,000 - 50,000. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water.Chemical Identifiers
| 9011-18-1 | |
| 40,000 | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
| (C6H7O5)m(C6H7O5)n | |
| MFCD00081551 | |
| Dextran sulfate sodium salt |
Specifications
| 9011-18-1 | |
| (C6H7O5)m(C6H7O5)n | |
| 25 g | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
| 40,000 | |
| 35-50% | |
| Poly Bottle |
| 100% | |
| MFCD00081551 | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| Dextran sulfate sodium salt | |
| Reagent | |
| 10% | |
| +75° to +105° |