missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran Sulfate Sodium Salt, Powder, Spectrum™ Chemical
DE136, 9011-18-1
Supplier: Spectrum Chemical Mfg Cor DE136100GM
Description
Spectrum™ Chemical Dextran Sulfate Sodium Salt, Powder is a sodium salt of the sulfuric acid ester of Dextran with an average molecular weight of 40,000 - 50,000. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water.Specifications
| 9011-18-1 | |
| (C6H7O5)m(C6H7O5)n | |
| 100 g | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
| 40,000 | |
| 35-50% | |
| Poly Bottle |
| 100% | |
| MFCD00081551 | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| Dextran sulfate sodium salt | |
| Reagent | |
| 10% | |
| +75° to +105° |
Chemical Identifiers
| 9011-18-1 | |
| 40,000 | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
| (C6H7O5)m(C6H7O5)n | |
| MFCD00081551 | |
| Dextran sulfate sodium salt |