missing translation for 'onlineSavingsMsg'
Learn More

Dextran, for GPC 5,000, MilliporeSigma™ Supelco™

Dextran is a polysaccharide. Dextran fluids behave as colloids; and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.

$489.84 - $489.84

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 9004-54-0
Physical Form Powder/Crystals
Linear Formula [C6H10O5 ]n
Technique GPC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015802
View Documents
Merck Emd Millipore
00269100MG
100 mg N/A
Only null left
 
Description

Description

Featured Industries: Food and Beverages
Specifications

Specifications

Bottomless Glass Bottle with Inserted Fused Cone
Powder/Crystals
GPC
Analytical Standard
9004-54-0
[C6H10O5 ]n
MFCD00130935
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'