missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Detoxi-Gel™ Endotoxin Removing Gel

Catalog No. pi20339
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
1000 mL
Catalog No. Quantity
PI20339 10 mL
PI20340 1000 mL
2 options

Catalog No. PI20339

Supplier: Thermo Scientific™ 20339

Only null left

Bind and extract endotoxins (lipopolysaccharides or LPS) from antibody or protein samples using bulk resin or columns of immobilized Polymixin B ligand.

Thermo Scientific™ Detoxi-Gel Endotoxin Removing Gel and Columns provide a quick method to remove endotoxins (lipopolysaccharides or LPS) from samples.

Detoxi-Gel Resin is a beaded agarose support onto which polymixin B has been immobilized. This ligand binds to the lipid A portion of lipopolysaccharides (LPS), which are the dominant form of endotoxins that affect biological samples. Even low levels of endotoxins can be toxic to cells or organisms and must be removed before biological samples can be added to cells for various treatment experiments. Detoxi-Gel Resin provides a quick method for removal of endotoxins from samples. It is available in convenient pre-packed columns and two sizes of slurry for use in small-scale or large-scale processes.

Highlights:

  • Affinity-based – uses immobilized polymixin B to remove endotoxins by binding to their lipid A domains

  • High capacity – 1mL of Detoxi-Gel Resin removes at least 9995 endotoxin units from a 5mL sample containing 10,000 EU of lipopolysaccharide (LPS)

  • Reusable – no loss in binding capacity even after 10 regenerations; requires activation and regeneration with 1% sodium deoxycholate (Part No. 89904)

  • Testing required – protein recovery is dependent upon protein type and concentration; empirical testing is required to validate the method for particular protein samples

  • Versatile – remove endotoxin from protein solutions, cell culture media and pharmacological components

Requires:

Sodium deoxycholate (Part No. 89904) for resin activation and regeneration.

Specifications

Capacity (Metric) 4000 to 6000 EU Binding per mL Gel
Content And Storage Store at 4°C.
Form Liquid
Product Line Detoxi-Gel
Quantity 10 mL
Target Endotoxin Removal
Type Endotoxin Removing Gel
Stationary Phase Polymixin B
Sufficient For 1 mL of gel binds 4000-6000 EU
Format Bottle
Column Type Affinity, Agarose Resin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.