missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Detergent Removal Spin Columns & Plates

Catalog No. PI87780
Change view
Click to view available options
Quantity:
2 x 96-well Plates
24 x 0.1 mL Columns
25 x 500 μL Columns
5 x 2 mL Columns
5 x 4 mL Columns
5 mL
10 mL
Product Line:
HiPPR
Pierce
Product Type:
Detergent Removal Resin
Spin Column
Spin Plate
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Line Product Type
PI87780 10 mL Pierce Detergent Removal Resin
PI87777 25 x 500 μL Columns Pierce Spin Column
PI87778 5 x 2 mL Columns Pierce Spin Column
PI87779 5 x 4 mL Columns Pierce Spin Column
PI88304 2 x 96-well Plates Pierce Spin Plate
PI88305 5 mL HiPPR Spin Column
PI88306 24 x 0.1 mL Columns HiPPR Spin Column
PI88307 2 x 96-well Plates HiPPR Spin Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. PI87780 Supplier Thermo Scientific™ Supplier No. 87780

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly eliminate high concentrations of detergents with minimal sample loss with detergent removal resins. Thermo Scientific HiPPR (High Protein and Peptide Recovery) resin is recommended for protein concentrations up to 100 μg/mL, while Thermo Scientific Pierce resin is suitable for concentrations over 100 μg/mL.

Detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing, and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis. Even low concentrations of detergents will contaminate instruments and interfere with column binding, elution, and peptide ionization. Thermo Scientific HiPPR and Pierce detergent removal resins are ideal for rapid removal of common detergents from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis.

  • Optimized—removes >95% of detergent from samples of proteins or peptides
  • Fast—sample processing takes less than 15 minutes
  • Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry

Pierce Detergent Removal Resin

Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.

HiPPR Detergent Removal Spin Column Kit

HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.

Available formats:

  • Resin slurries—bulk resin supplied with empty 0.8-mL spin columns
  • Spin columns—0.1 mL micro-centrifuge columns
  • 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification

Specifications

Description Bottle
Product Type Detergent Removal Resin
Content And Storage Upon receipt store at 4°C. The resin is stored in 0.15 M NaCl, 0.05% sodium azide.
Column Type Proprietary Resin, Affinity
Stationary Phase Proprietary Ligand
Form Liquid
Product Line Pierce
Purification Target Detergent Removal
Sufficient For 1 to 2.5 mL of Typical Detergent-Containing Sample
Quantity 10 mL
Format Bottle
Show More Show Less
I still have detergent present in my sample after protein digestion. Is there anything I can do?

Some detergents can be removed from peptide samples using Pierce Detergent Removal Resin (Cat. No. 87780) or HiPPR Detergent Removal Spin Columns (Cat. No. 88306) in the HIPPR (High Protein and Peptide Recovery) format. However, detergents are more easily removed at the protein level using acetone precipitation, dialysis, or affinity capture using Pierce Detergent Removal Resin and HiPPR Detergent Removal Spin Columns before protein digestion.

What is the best way to remove detergents from my sample for mass spectrometry analysis?

Detergents are more easily removed at the protein level using acetone precipitation (https://tools.thermofisher.com/content/sfs/brochures/TR0049-Acetone-precipitation.pdf), dialysis, or affinity capture using Pierce Detergent Removal Resin (Cat. No. 87780) or HiPPR Detergent Removal Spin Columns (Cat. No. 88306).