missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmoplakin Recombinant Rabbit Monoclonal Antibody (23GB5220)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | Desmoplakin |
|---|---|
| Clone | 23GB5220 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
Description
Desmoplakin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.Specifications
| Desmoplakin | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| DSP | |
| 2300002E22Rik; 250/210 kDa paraneoplastic pemphigus antigen; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; dermatan sulfate proteoglycan 3; desmoplakin; desmoplakin I/II; Desmoplakin II; desmoplakin; LOW QUALITY PROTEIN: desmoplakin; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP; Dsp; dspg3; Oculoglycan; Opt; Optc; optcl; Opticin; rul; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 | |
| DSP | |
| Primary | |
| Affinity chromatography |
| 23GB5220 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15924 | |
| 1832 | |
| A synthesized peptide derived from human Desmoplakin (400-470AA). | |
| Antibody |