missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Desmoplakin Recombinant Rabbit Monoclonal Antibody (23GB5220)

Catalog No. MA553869
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553869 20 μL
MA553868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553869 Supplier Invitrogen™ Supplier No. MA553869
Only null left

Rabbit Recombinant Monoclonal Antibody

Desmoplakin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.

Specifications

Antigen Desmoplakin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5220
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DSP
Gene Accession No. P15924
Gene Alias 2300002E22Rik; 250/210 kDa paraneoplastic pemphigus antigen; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; dermatan sulfate proteoglycan 3; desmoplakin; desmoplakin I/II; Desmoplakin II; desmoplakin; LOW QUALITY PROTEIN: desmoplakin; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP; Dsp; dspg3; Oculoglycan; Opt; Optc; optcl; Opticin; rul; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047
Gene Symbols DSP
Host Species Rabbit
Immunogen A synthesized peptide derived from human Desmoplakin (400-470AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1832
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less