missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Deoxycholic Acid, Sodium Salt

Catalog No. ICN10290690
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10290690 500 g
ICN10290625 25 g
ICN10290680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10290690 Supplier MP Biomedicals™ Supplier No. 0210290690

missing translation for 'validMainframeMsg'

Deoxycholic Acid, Sodium Salt is an anionic detergent used to solubilize membrane-bound proteins.

  • Deoxycholic acid, sodium salt is an anionic detergent useful for the extraction of membrane receptors and other plasma membrane proteins and for nuclei isolation.
  • Deoxycholic acid has been used in a modified procedure to recover 40 to 80% of a protein from a 1 μg/mL solution.

Specifications

Color White
Product Type Detergent
CAS 302-95-4
Molecular Formula C24H39NaO4
Formula Weight 414.55
Form Powder
Quantity 500 g

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.