missing translation for 'onlineSavingsMsg'
Learn More

Deoxy-bigCHAP

Analog of CHAPS and CHAPSO | CAS: 86303-23-3 | C42H75N3O15 | 862.068 g/mol

Supplier:  Thermo Scientific Chemicals J6457803

 View more versions of this product

Catalog No. AAJ6457803


Only null left

Description

Description

Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.

Solubility
Soluble in water at 50mg/ml. Also soluble in DMSO or methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Deoxy-bigCHAP
86303-23-3
1 g
MFCD00161482
N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide
OJSUWTDDXLCUFR-HGZMBBKESA-N
(2R,3S,4R,5R)-N-[3-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propyl]amino
446320
White
Powder
C42H75N3O15
14,1214
Soluble in water at 50mg/ml; Also soluble in DMSO or methanol.
CC(CCC(=O)N(CCCNC(=O)C(C(C(C(CO)O)O)O)O)CCCNC(=O)C(C(C(C(CO)O)O)O)O)C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C
862.068
862.05

Chemical Identifiers

86303-23-3
862.068
OJSUWTDDXLCUFR-HGZMBBKESA-N
446320
CC(CCC(=O)N(CCCNC(=O)C(C(C(C(CO)O)O)O)O)CCCNC(=O)C(C(C(C(CO)O)O)O)O)C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C
C42H75N3O15
MFCD00161482
N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide
(2R,3S,4R,5R)-N-[3-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propyl]amino
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only