missing translation for 'onlineSavingsMsg'
Learn More

Deoxy-bigCHAP

Catalog No. p-7042954
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64578MD 250 mg
AAJ6457803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64578MD Supplier Thermo Scientific Chemicals Supplier No. J64578MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analog of CHAPS and CHAPSO | CAS: 86303-23-3 | C42H75N3O15 | 862.068 g/mol

Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.

Solubility
Soluble in water at 50mg/ml. Also soluble in DMSO or methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 86303-23-3
Molecular Formula C42H75N3O15
Molecular Weight (g/mol) 862.068
MDL Number MFCD00161482
InChI Key OJSUWTDDXLCUFR-HGZMBBKESA-N
Synonym N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide
PubChem CID 446320
IUPAC Name (2R,3S,4R,5R)-N-[3-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propyl]amino
SMILES CC(CCC(=O)N(CCCNC(=O)C(C(C(C(CO)O)O)O)O)CCCNC(=O)C(C(C(C(CO)O)O)O)O)C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C

Specifications

Color White
Physical Form Powder
Quantity 250 mg
Merck Index 14,1214
Solubility Information Soluble in water at 50mg/ml; Also soluble in DMSO or methanol.
Formula Weight 862.05
Chemical Name or Material Deoxy-bigCHAP
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only