missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydroacetic Acid 98.0+%, TCI America™
$46.60 - $171.49
Chemical Identifiers
| CAS | 520-45-6 |
|---|---|
| Molecular Formula | C8H8O4 |
| Molecular Weight (g/mol) | 168.148 |
| MDL Number | MFCD00066709 |
| InChI Key | PGRHXDWITVMQBC-UHFFFAOYSA-N |
| Synonym | dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4 |
| PubChem CID | 122903 |
| IUPAC Name | 3-acetyl-6-methylpyran-2,4-dione |
| SMILES | CC1=CC(=O)C(C(=O)O1)C(=O)C |
Chemical Identifiers
| 520-45-6 | |
| 168.148 | |
| PGRHXDWITVMQBC-UHFFFAOYSA-N | |
| 122903 | |
| CC1=CC(=O)C(C(=O)O1)C(=O)C |
| C8H8O4 | |
| MFCD00066709 | |
| dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4 | |
| 3-acetyl-6-methylpyran-2,4-dione |
Specifications
| 520-45-6 | |
| 270°C | |
| MFCD00066709 | |
| dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4 | |
| CC1=CC(=O)C(C(=O)O1)C(=O)C | |
| 168.148 | |
| 168.15 | |
| Crystalline Powder |
| 111°C | |
| C8H8O4 | |
| 25 g | |
| PGRHXDWITVMQBC-UHFFFAOYSA-N | |
| 3-acetyl-6-methylpyran-2,4-dione | |
| 122903 | |
| ≥98.0% (GC,T) | |
| Dehydroacetic Acid |
Safety and Handling
EINECSNumber : (5)-0268
RTECSNumber : UP8050000
TSCA : Yes