missing translation for 'onlineSavingsMsg'
Learn More

Dehydroacetic Acid 98.0+%, TCI America™

Supplier:  TCI America D0039500G

 View more versions of this product

Catalog No. D0039500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dehydroacetic Acid
111°C
C8H8O4
500 g
PGRHXDWITVMQBC-UHFFFAOYSA-N
3-acetyl-6-methylpyran-2,4-dione
122903
≥98.0% (GC,T)
520-45-6
270°C
MFCD00066709
dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
CC1=CC(=O)C(C(=O)O1)C(=O)C
168.148
168.15
Crystalline Powder

Chemical Identifiers

520-45-6
168.148
PGRHXDWITVMQBC-UHFFFAOYSA-N
122903
CC1=CC(=O)C(C(=O)O1)C(=O)C
C8H8O4
MFCD00066709
dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
3-acetyl-6-methylpyran-2,4-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0268

RTECSNumber : UP8050000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions