missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ 96-Well Deep Well Plates, Square V-Bottom
BIO-CERT™ PCR QUALITY deep well plates are manufactured under cleanroom conditions and designed for a wide range of applications.
$1,738.53 - $2,207.28
Specifications
| Barcode | No Barcode |
|---|---|
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
Description
- V-bottoms for maximum sample recovery
- Available sterile or non-sterile
- Virgin PP plates with a 12 x 8 configuration
- 2.2 mL nominal volume
- Matching sterile and non-sterile sealing mats are available separately
Specifications
| No Barcode | |
| Without Lid | |
| Autoclavable | |
| BRAND | |
| Single-use | |
| Multi-channel Pipettes, LH Robots, Plate Handlers | |
| Polypropylene | |
| 96 | |
| Non-treated | |
| Yes | |
| SBS/ANSI | |
| Yes | |
| 127.76 mm | |
| 44.20 mm |
| Standard | |
| 8 x 12 | |
| Untreated | |
| Natural | |
| Deepwell Plates | |
| Full | |
| 2.2 mL | |
| 2.2 mL | |
| Square, Conical Bottom (V-well) | |
| HTS, cell and tissue cultures, serial dilutions, reagent transfers, and sample storage | |
| 50 Pk. | |
| Natural | |
| 85.48 mm |