missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ 96-Well Deep Well Plates, Square V-Bottom

Catalog No. 14380946
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14380946 Non-sterile
14380947 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14380946 Supplier BRANDTECH™ BRAND™ Supplier No. DW2200

BIO-CERT™ PCR QUALITY deep well plates are manufactured under cleanroom conditions and designed for a wide range of applications.

  • V-bottoms for maximum sample recovery
  • Available sterile or non-sterile
  • Virgin PP plates with a 12 x 8 configuration
  • 2.2 mL nominal volume
  • Matching sterile and non-sterile sealing mats are available separately

Specifications

Barcode No Barcode
Format Standard
Lid Without Lid
Description Deep-well plate, 2.2mL square wells with conical bottom, pack of 50
Array 8 x 12
Autoclavable Autoclavable
Binding Type Untreated
Product Line BRAND
Color Natural
Disposable Single-use
Product Type Deepwell Plates
For Use With (Equipment) Multi-channel Pipettes, LH Robots, Plate Handlers
Skirt Style Full
Material Polypropylene
Volume (Metric) Well 2.2 mL
No. of Wells 96
Volume (Metric) Working 2.2 mL
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 212.2 mL
Well Shape Square, Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) HTS, cell and tissue cultures, serial dilutions, reagent transfers, and sample storage
Plate Compatibility SBS/ANSI
Quantity 50 Pk.
RNase Free Yes
Well Color Natural
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 44.20 mm
Show More Show Less