missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DDMAB, ≥98%, Calbiochem™,

Catalog No. 2520005GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2520005GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2520005GM Supplier MilliporeSigma™ Supplier No. 2520005GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water-soluble zwitterionic detergent efficient for the extraction of mycoplasma membrane antigens under mild conditions.

Chemical Identifiers

CAS 15163-30-1

Specifications

Color White
Quantity 5 g
Format Glass Bottle