missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dansylglycine 98.0+%, TCI America™
$59.69 - $527.49
Chemical Identifiers
| CAS | 1091-85-6 |
|---|---|
| Molecular Formula | C14H16N2O4S |
| Molecular Weight (g/mol) | 308.352 |
| MDL Number | MFCD00037734 |
| InChI Key | QHFXORCWAQTTGH-UHFFFAOYSA-N |
| Synonym | N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, Dns-Gly-OH |
| PubChem CID | 70666 |
| IUPAC Name | 2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]acetic acid |
| SMILES | CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D1501100MG
|
TCI America
D1501100MG |
100 mg |
N/A
|
|
|||||
|
D15011G
|
TCI America
D15011G |
1 g |
N/A
|
|
|||||
|
D15015G
|
TCI America
D15015G |
5 g |
N/A
|
|
|||||
Chemical Identifiers
| 1091-85-6 | |
| 308.352 | |
| QHFXORCWAQTTGH-UHFFFAOYSA-N | |
| 70666 | |
| CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NCC(=O)O |
| C14H16N2O4S | |
| MFCD00037734 | |
| N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, Dns-Gly-OH | |
| 2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]acetic acid |
Specifications
| 159°C | |
| C14H16N2O4S | |
| N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, Dns-Gly-OH | |
| CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NCC(=O)O | |
| 100 mg | |
| 70666 | |
| ≥98.0% (T) | |
| Dansylglycine |
| 1091-85-6 | |
| MFCD00037734 | |
| QHFXORCWAQTTGH-UHFFFAOYSA-N | |
| 2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]acetic acid | |
| 308.352 | |
| 308.35 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes