missing translation for 'onlineSavingsMsg'
Learn More

Dansylglycine 98.0+%, TCI America™

Catalog No. p-7128761
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D540625MG 25 mg
D5406100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D540625MG Supplier TCI America Supplier No. D540625MG

Chemical Identifiers

CAS 1091-85-6
Molecular Formula C14H16N2O4S
Molecular Weight (g/mol) 308.352
MDL Number MFCD00037734
InChI Key QHFXORCWAQTTGH-UHFFFAOYSA-N
Synonym N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, Dns-Gly-OH
PubChem CID 70666
IUPAC Name 2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]acetic acid
SMILES CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NCC(=O)O

Specifications

Melting Point 159°C
Quantity 25 mg
Formula Weight 308.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansylglycine [for Albumin binding assay]