missing translation for 'onlineSavingsMsg'
Learn More

Dansyl-L-valine 98.0+%, TCI America™

Catalog No. d15051g
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15051G 1 g
D1505100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15051G Supplier TCI America Supplier No. D15051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1098-50-6
Molecular Formula C17H22N2O4S
Molecular Weight (g/mol) 350.433
MDL Number MFCD00069524
InChI Key VULDIKGSZAUMHB-INIZCTEOSA-N
Synonym 5-Dimethylaminonaphthalene-1-sulfonyl-L-valine, Dns-Val-OH
PubChem CID 13734194
IUPAC Name (2S)-2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)NS(=O)(=O)C1=CC=CC2=C1C=CC=C2N(C)C

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 1 g
Formula Weight 350.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansyl-L-valine