accessibility menu, dialog, popup

UserName

H3CCH3OHONHSOONCH3H3C

CAS RN 1098-50-6

IUPAC Name: 2-[5-(dimethylamino)naphthalene-1-sulfonamido]-3-methylbutanoic acid
Synonyms: 2-[5-(dimethylamino)naphthalene-1-sulfonamido]-3-methylbutanoic acid
Molecular Weight (g/mol): 350.43
Molecular Formula: C17H22N2O4S
InChi Key: VULDIKGSZAUMHB-UHFFFAOYNA-N
SMILES: CC(C)C(NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results