missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D3710-95 Calibration Mix, Restek™
Description
1mL/ampule contains
- n-pentane (C5) 8%
- n-hexane (C6) 6%
- n-heptane (C7) 10%
- n-octane (C8) 5%
- n-decane (C10) 4%
- n-dodecane (C12) 4%
- n-tridecane (C13) 2%
- n-tetradecane (C14) 2%
- n-pentadecane (C15) 2%
- 2-methylbutane 10%
- 2-methylpentane 6%
- 2,4-dimethylpentane 6%
- toluene 12%
- p-xylene 14%
- n-propylbenzene 5%
- n-butylbenzene 4%
Specifications
Specifications
| Packaging | Ampule |
| Type | Multi-component Standard |
| Quantity | 1 mL |