missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Valine 98.0+%, TCI America™
$47.46 - $265.80
Chemical Identifiers
| CAS | 640-68-6 |
|---|---|
| Molecular Formula | C5H11NO2 |
| Molecular Weight (g/mol) | 117.15 |
| MDL Number | MFCD00064219 |
| InChI Key | KZSNJWFQEVHDMF-SCSAIBSYSA-N |
| Synonym | d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid |
| PubChem CID | 71563 |
| ChEBI | CHEBI:27477 |
| IUPAC Name | (2R)-2-amino-3-methylbutanoic acid |
| SMILES | CC(C)[C@@H](N)C(O)=O |
Chemical Identifiers
| C5H11NO2 | |
| MFCD00064219 | |
| d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid | |
| CHEBI:27477 | |
| CC(C)[C@@H](N)C(O)=O |
Specifications
| 640-68-6 | |
| C5H11NO2 | |
| d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid | |
| CC(C)[C@@H](N)C(O)=O | |
| 117.15 | |
| CHEBI:27477 | |
| ≥98.0% (T) | |
| D-Valine |
| White | |
| MFCD00064219 | |
| KZSNJWFQEVHDMF-SCSAIBSYSA-N | |
| (2R)-2-amino-3-methylbutanoic acid | |
| 71563 | |
| 117.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1604
RTECSNumber : YV9360000
TSCA : Yes