missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Valine, 98+%
CAS: 640-68-6 | C5H11NO2 | 117.15 g/mol
$69.16 - $237.41
Chemical Identifiers
| CAS | 640-68-6 |
|---|---|
| Molecular Formula | C5H11NO2 |
| Molecular Weight (g/mol) | 117.15 |
| MDL Number | MFCD00064219 |
| InChI Key | KZSNJWFQEVHDMF-SCSAIBSYSA-N |
| Synonym | d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid |
| PubChem CID | 71563 |
| ChEBI | CHEBI:27477 |
| IUPAC Name | (2R)-2-amino-3-methylbutanoic acid |
| SMILES | CC(C)[C@@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC227220050
|
Thermo Scientific Chemicals
227220050 |
5 g | Glass Bottle |
Each for $69.16
|
|
||||
|
AC227220250
|
Thermo Scientific Chemicals
227220250 |
25 g | Glass Bottle |
Each for $237.41
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C5H11NO2 | |
| MFCD00064219 | |
| d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid | |
| CHEBI:27477 | |
| CC(C)[C@@H](N)C(O)=O |
Specifications
| 302.0°C to 303.0°C | |
| White | |
| 0.5% max. (105°C, 4 hrs) | |
| C5H11NO2 | |
| MFCD00064219 | |
| d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid | |
| Solubility in water: 56g/L soluble in water (20°C). Other solubilities: soluble in mineral acids, insoluble in organic solvents, soluble in ethanol, diethyl ether, benzene | |
| KZSNJWFQEVHDMF-SCSAIBSYSA-N | |
| −27.50 (20.00°C c=5,5N HCl) | |
| 117.15 | |
| CHEBI:27477 | |
| ≥98% | |
| D-Valine, 98+% |
| 640-68-6 | |
| Authentic | |
| 98+% | |
| (CH3)2CHCH(NH2)CO2H | |
| − 27.50 | |
| 0.2% max. | |
| 0.5% max. | |
| CC(C)[C@@H](N)C(O)=O | |
| (2R)-2-amino-3-methylbutanoic acid | |
| 71563 | |
| 117.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : 211-368-9
RTECSNumber : YV9360000
TSCA : TSCA
RUO – Research Use Only