missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ D-Tube™ Dialyzer Mini, MWCO 6-8 kDa

Catalog No. 715043
Change view
Click to view available options
Quantity:
10d-tu
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
715043 10d-tu
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 715043 Supplier MilliporeSigma™ Supplier No. 715043

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for dialysis and electroelution of proteins, RNA, DNA, and oligonucleotides from polyacrylamide or agarose gels

  • Easy-to-handle dialyzers for buffer exchange and removal of urea, detergents
  • Ideal for electroelution of proteins, protein-DNA complexes, oligonucleotides, DNA, and RNA from polyacrylamide and agarose gels
  • One-step procedure that does not require syringes or any special equipment
  • Sample volume recovery > 97%
  • Protease, RNAse, DNAse, and PCR product free

Specifications

Product Type D-Tube Dialyzer Mini, MWCO 6-8 kDa
Quantity 10d-tu
Includes
  • 10 D-Tube Dialyzers and one floating rack that can hold up to four D-Tube Dialyzers
  • The D-Tube Dialyzers Mega contain 10 or 50 tubes and one or 5 floating racks to hold one D-Tube Mega in the exchange buffer
Content And Storage +15°C to +30°C
For Use With (Application)
  • MALDIMS, animal immunization for antibody production, HPLC analysis, peptide mapping, and functional assays
  • In addition, D-Tube Dialyzers can be used for oligonucleotides, RNA and DNA extraction from both polyacrylamide and agarose gels