missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ D-Tube™ Dialyzer Mega, 15mL, MWCO 6-8 kDa
Description
- Easy-to-handle dialyzers for buffer exchange and removal of urea, detergents
- Ideal for electroelution of proteins, protein-DNA complexes, oligonucleotides, DNA, and RNA from polyacrylamide and agarose gels
- One-step procedure that does not require syringes or any special equipment
- Sample volume recovery > 97%
- Protease, RNAse, DNAse, and PCR product free
Specifications
Specifications
| Product Type | D-Tube Dialyzer Mega, 15mL, MWCO 6-8 kDa |
| Quantity | 10d-tu |
| Includes | 10 or 50 tubes and one or 5 floating racks to hold one D-Tube Mega in the exchange buffer |
| Content And Storage | +15°C to +30°C |