missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ D-Tube™ Dialyzer Mega, 10mL, MWCO 6-8 kDa

Catalog No. p-6810183
Change view
Click to view available options
Quantity:
10d-tu
50d-tu
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
717403 10d-tu
717404 50d-tu
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 717403 Supplier MilliporeSigma™ Supplier No. 717403
Only null left

Used for dialysis and electroelution of proteins, RNA, DNA, and oligonucleotides from polyacrylamide or agarose gels

  • Easy-to-handle dialyzers for buffer exchange and removal of urea, detergents
  • Ideal for electroelution of proteins, protein-DNA complexes, oligonucleotides, DNA, and RNA from polyacrylamide and agarose gels
  • One-step procedure that does not require syringes or any special equipment
  • Sample volume recovery > 97%
  • Protease, RNAse, DNAse, and PCR product free

Specifications

Product Type D-Tube Dialyzer Mega, 10mL, MWCO 6-8 kDa
Quantity 10d-tu
Includes 10 or 50 tubes and one or 5 floating racks to hold one D-Tube Mega in the exchange buffer
Content And Storage +15™C to +30™C