missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Trehalose, Dihydrate, Spectrum™ Chemical
T1097, 6138-23-4, C12H22O11·2H2O
$212.33 - $56,382.06
Chemical Identifiers
| CAS | 6138-23-4 |
|---|---|
| Molecular Formula | C12H26O13 |
| Molecular Weight (g/mol) | 378.33 |
| MDL Number | MFCD00071594 |
| InChI Key | DPVHGFAJLZWDOC-DJCWUSJTNA-N |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate |
| SMILES | O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18601998
|
N/A | 5 g | Amber Glass Bottle |
Each for $212.33
|
|
||||
|
18601999
|
N/A | 25 g | Poly Bottle |
Each for $437.64
|
|
||||
|
18602000
|
N/A | 100 g | Poly Bottle |
Each for $1,048.36
|
|
||||
|
18602001
|
N/A | 500 g | Poly Bottle |
Each for $3,541.33
|
|
||||
|
18602002
|
N/A | 1 kg | Poly Pail |
Each for $8,956.03
|
|
||||
|
18602003
|
N/A | 5 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602004
|
N/A | 50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical D-(+)-Trehalose, Dihydrate , is a natural alpha-linked disaccharide that is also referred to as mycose or tremalose. It is found naturally in plants and microorganisms such as grasshoppers, shrimp, butterflies and bees. It is thought to have a use in the treatment for Huntington′s and Parkinson′s disease as it possibly corrects defects in autophagy seen in these diseases.Chemical Identifiers
| 6138-23-4 | |
| 378.33 | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| C12H26O13 | |
| MFCD00071594 | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate |
Specifications
| 6138-23-4 | |
| 0.05% | |
| MFCD00071594 | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate | |
| Reagent |
| 100% | |
| C12H26O13 | |
| +197° to +201° | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
| 378.33 |