missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Trehalose, Dihydrate, Spectrum™ Chemical

T1097, 6138-23-4, C12H22O11·2H2O

Supplier:  Spectrum Chemical Mfg Cor T109725GM

Quantity 25 g
Packaging Poly Bottle

 View more versions of this product

Catalog No. 18601999


Only null left

Description

Description

Spectrum™ Chemical D-(+)-Trehalose, Dihydrate , is a natural alpha-linked disaccharide that is also referred to as mycose or tremalose. It is found naturally in plants and microorganisms such as grasshoppers, shrimp, butterflies and bees. It is thought to have a use in the treatment for Huntington′s and Parkinson′s disease as it possibly corrects defects in autophagy seen in these diseases.
Specifications

Chemical Identifiers

6138-23-4
378.33
DPVHGFAJLZWDOC-DJCWUSJTNA-N
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
C12H26O13
MFCD00071594
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate

Specifications

6138-23-4
0.05%
C12H26O13
25 g
DPVHGFAJLZWDOC-DJCWUSJTNA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
Reagent
100%
Poly Bottle
MFCD00071594
+197° to +201°
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
378.33
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions