missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| D-Sorbitol | |
| 110°C | |
| 200°C | |
| MFCD00004708 | |
| d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol | |
| C(C(C(C(C(CO)O)O)O)O)O | |
| 182.172 | |
| CHEBI:17924 | |
| ≥97.0% (GC) |
| 50-70-4 | |
| White | |
| C6H14O6 | |
| 500 g | |
| FBPFZTCFMRRESA-JGWLITMVSA-N | |
| (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol | |
| 5780 | |
| 182.17 | |
| Crystalline Powder |
Chemical Identifiers
| 50-70-4 | |
| 182.172 | |
| FBPFZTCFMRRESA-JGWLITMVSA-N | |
| 5780 | |
| (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol |
| C6H14O6 | |
| MFCD00004708 | |
| d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol | |
| CHEBI:17924 | |
| C(C(C(C(C(CO)O)O)O)O)O |
Safety and Handling
EINECSNumber : (8)-0050
RTECSNumber : LZ4290000
TSCA : Yes