missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol 97.0+%, TCI America™

Catalog No. S006525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S006525G 25 g
S0065500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S006525G Supplier TCI America Supplier No. S006525G

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00004708
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 110°C
Color White
Boiling Point 200°C
Quantity 25 g
Formula Weight 182.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-Sorbitol
EINECSNumber (8)-0050
RTECSNumber LZ4290000
TSCA Yes