missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Ribose 98.0+%, TCI America™

Catalog No. p-7135666
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R002525G 25 g
R0025250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R002525G Supplier TCI America Supplier No. R002525G
Only null left

Chemical Identifiers

CAS 50-69-1
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135453
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917
PubChem CID 5311110
ChEBI CHEBI:47014
IUPAC Name (3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
SMILES OC[C@H]1OC(O)[C@H](O)[C@@H]1O

Specifications

Color White
Quantity 25 g
Formula Weight 150.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Ribose
EINECSNumber (8)-0048
RTECSNumber VJ2275000
TSCA Yes