missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Raffinose pentahydrate, 99%

CAS: 17629-30-0 | C18H42O21 | 594.513 g/mol

Supplier:  Thermo Scientific Chemicals A1831309

 View more versions of this product

Catalog No. AAA1831309


Only null left
Explore available promotions

Description

Description

D-(+)-Raffinose pentahydrate is used as a carbon source in yeast culture media. It is also used in moisturizers, prevent skin ageing, promotes skin regeneration, additive in low-cariogenic food and drinks, non-digestible with lower efficiency than lactose or sucrose and stimulates the growth of lactobacilli and bifidobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Raffinose pentahydrate is used as a carbon source in yeast culture media. It is also used in moisturizers, prevent skin ageing, promotes skin regeneration, additive in low-cariogenic food and drinks, non-digestible with lower efficiency than lactose or sucrose and stimulates the growth of lactobacilli and bifidobacteria.

Solubility
Soluble in pyridine and water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

D-(+)-Raffinose pentahydrate
78°C to 82°C
C18H42O21
10 g
Hygroscopic
Soluble in pyridine and water.
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O
594.513
594.52 (504.45 Anhydrous)
17629-30-0
0.99
MFCD00071590
4285763
d-+-raffinosepentahydrate
BITMAWRCWSHCRW-ORBWWOJFSA-N
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
134129414

Chemical Identifiers

17629-30-0
594.513
BITMAWRCWSHCRW-ORBWWOJFSA-N
134129414
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O
C18H42O21
MFCD00071590
d-+-raffinosepentahydrate
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-146-9

RTECSNumber : LZ5851200

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only