missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Raffinose pentahydrate, 99%

Catalog No. p-3173068
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1831309 10 g
AAA1831318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1831309 Supplier Thermo Scientific Chemicals Supplier No. A1831309

CAS: 17629-30-0 | C18H42O21 | 594.513 g/mol

D-(+)-Raffinose pentahydrate is used as a carbon source in yeast culture media. It is also used in moisturizers, prevent skin ageing, promotes skin regeneration, additive in low-cariogenic food and drinks, non-digestible with lower efficiency than lactose or sucrose and stimulates the growth of lactobacilli and bifidobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Raffinose pentahydrate is used as a carbon source in yeast culture media. It is also used in moisturizers, prevent skin ageing, promotes skin regeneration, additive in low-cariogenic food and drinks, non-digestible with lower efficiency than lactose or sucrose and stimulates the growth of lactobacilli and bifidobacteria.

Solubility
Soluble in pyridine and water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H42O21
Molecular Weight (g/mol) 594.513
MDL Number MFCD00071590
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Melting Point 78°C to 82°C
Assay Percent Range 0.99
Quantity 10 g
Beilstein 4285763
Sensitivity Hygroscopic
Solubility Information Soluble in pyridine and water.
Formula Weight 594.52 (504.45 Anhydrous)
Chemical Name or Material D-(+)-Raffinose pentahydrate
EINECSNumber 208-146-9
RTECSNumber LZ5851200
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only