missing translation for 'onlineSavingsMsg'
Learn More

D-Norleucine 98.0+%, TCI America™

Supplier:  TCI America N03271G

 View more versions of this product

Catalog No. N03271G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

D-Norleucine
White
MFCD00008099
d-norleucine, h-d-nle-oh, r-2-aminohexanoic acid, d-2-aminohexanoic acid, d---norleucine, 2r-2-aminohexanoic acid, unii-vp8g7lx265, norleucine, norleucine, d--, d-nor-leucine
CCCC[C@@H](N)C(O)=O
131.18
CHEBI:42101
≥98.0% (HPLC,T)
327-56-0
C6H13NO2
1 g
LRQKBLKVPFOOQJ-RXMQYKEDSA-N
(2R)-2-aminohexanoic acid
456468
131.18
Crystalline Powder

Chemical Identifiers

327-56-0
131.18
LRQKBLKVPFOOQJ-RXMQYKEDSA-N
456468
(2R)-2-aminohexanoic acid
C6H13NO2
MFCD00008099
d-norleucine, h-d-nle-oh, r-2-aminohexanoic acid, d-2-aminohexanoic acid, d---norleucine, 2r-2-aminohexanoic acid, unii-vp8g7lx265, norleucine, norleucine, d--, d-nor-leucine
CHEBI:42101
CCCC[C@@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions