missing translation for 'onlineSavingsMsg'
Learn More

D-Norleucine 98.0+%, TCI America™

Catalog No. p-7133751
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03271G 1 g
N032710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03271G Supplier TCI America Supplier No. N03271G
Only null left

Chemical Identifiers

CAS 327-56-0
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00008099
InChI Key LRQKBLKVPFOOQJ-RXMQYKEDSA-N
Synonym d-norleucine, h-d-nle-oh, r-2-aminohexanoic acid, d-2-aminohexanoic acid, d---norleucine, 2r-2-aminohexanoic acid, unii-vp8g7lx265, norleucine, norleucine, d--, d-nor-leucine
PubChem CID 456468
ChEBI CHEBI:42101
IUPAC Name (2R)-2-aminohexanoic acid
SMILES CCCC[C@@H](N)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 131.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Norleucine