accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 327-56-0

H3C(R)NH2OHO

CAS RN 327-56-0

IUPAC Name: (2R)-2-aminohexanoic acid
Synonyms: d-norleucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: LRQKBLKVPFOOQJ-RXMQYKEDSA-N
SMILES: CCCC[C@@H](N)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.