missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| D-Norleucine | |
| White | |
| MFCD00008099 | |
| d-norleucine, h-d-nle-oh, r-2-aminohexanoic acid, d-2-aminohexanoic acid, d---norleucine, 2r-2-aminohexanoic acid, unii-vp8g7lx265, norleucine, norleucine, d--, d-nor-leucine | |
| CCCC[C@@H](N)C(O)=O | |
| 131.18 | |
| CHEBI:42101 | |
| ≥98.0% (HPLC,T) |
| 327-56-0 | |
| C6H13NO2 | |
| 10 g | |
| LRQKBLKVPFOOQJ-RXMQYKEDSA-N | |
| (2R)-2-aminohexanoic acid | |
| 456468 | |
| 131.18 | |
| Crystalline Powder |
Chemical Identifiers
| 327-56-0 | |
| 131.18 | |
| LRQKBLKVPFOOQJ-RXMQYKEDSA-N | |
| 456468 | |
| (2R)-2-aminohexanoic acid |
| C6H13NO2 | |
| MFCD00008099 | |
| d-norleucine, h-d-nle-oh, r-2-aminohexanoic acid, d-2-aminohexanoic acid, d---norleucine, 2r-2-aminohexanoic acid, unii-vp8g7lx265, norleucine, norleucine, d--, d-nor-leucine | |
| CHEBI:42101 | |
| CCCC[C@@H](N)C(O)=O |
Safety and Handling
TSCA : No