missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Myo-inositol, 99.26%, For HPLC analysis, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210205280
Specifications
| D-Myo-inositol | |
| 87-89-8 | |
| 6.2g/mL | |
| C6H12O6 | |
| scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
| C1(C(C(C(C(C1O)O)O)O)O)O | |
| 180.156 | |
| CHEBI:24848 | |
| Powder |
| For HPLC analysis | |
| White | |
| 99.26% | |
| 100 g | |
| CDAISMWEOUEBRE-UHFFFAOYSA-N | |
| cyclohexane-1,2,3,4,5,6-hexol | |
| 892 | |
| 180.2 |
Chemical Identifiers
| 87-89-8 | |
| 180.156 | |
| scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
| CHEBI:24848 | |
| C1(C(C(C(C(C1O)O)O)O)O)O |
| C6H12O6 | |
| CDAISMWEOUEBRE-UHFFFAOYSA-N | |
| 892 | |
| cyclohexane-1,2,3,4,5,6-hexol |